Learn More
Sigma Organic Chemistry 6-Chloro-3-hydroxypyrid | 1G | 19064-67-6 | MFCD00233955
Supplier: Sigma Organic Chemistry SYX002711G

6-Chloro-3-hydroxypyrid , 1G
About this item:
CAS #: 19064-67-6
MDL #: MFCD00233955
Chemical Formula: C4H3ClN2O
Molecular Weight: 130.53
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.