Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloro-3-indolyl-beta-D-glucuronide cyclohexylammonium salt, 98%, Thermo Scientific™

Catalog No. AAB21068MF
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB21068MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB21068MF Supplier Thermo Scientific Chemicals Supplier No. B21068MF

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138182-20-4
Molecular Formula C14H13ClNO7
Molecular Weight (g/mol) 342.71
MDL Number MFCD00153940
InChI Key UFBPRKNLSYGHJJ-BYNIDDHOSA-M
Synonym cyclohexanaminium 2s,3s,4s,5r,6s-6-6-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxytetrahydro-2h-pyran-2-carboxylate, salmon-glca, cyclohexylammonium ion 2s,3s,4s,5r,6s-6-6-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxyoxane-2-carboxylate, c14h14clno7.c6h13n, 6-chloro-3-indolyl b-d-glucuronide cyclohexyl, 6-chloro-3-indolyl beta-d-glucuronide cyclohexylammonium salt, 6-chloro-3-indoxyl-beta-d-glucuronic acid, cyclohexylammonium salt
PubChem CID 16211694
IUPAC Name (2S,3S,4S,5R,6S)-6-[(6-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylic acid;cyclohexanamine
SMILES O[C@@H]1[C@@H](O)[C@H](OC2=CNC3=CC(Cl)=CC=C23)O[C@@H]([C@H]1O)C([O-])=O

Specifications

Quantity 50 mg
Sensitivity Light and moisture sensitive
Formula Weight 442.9
Percent Purity 98%
Chemical Name or Material 6-Chloro-3-indolyl-beta-D-glucuronide cyclohexylammonium salt
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.