Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloro-4-hydroxycoumarin 97.0+%, TCI America™
SDP

Catalog No. C28701G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28701G 1 g
C28705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28701G Supplier TCI America Supplier No. C28701G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 19484-57-2
Molecular Formula C9H5ClO3
Molecular Weight (g/mol) 196.59
MDL Number MFCD00239399
InChI Key UJHIADILZZWIED-UHFFFAOYSA-N
Synonym 6-chloro-4-hydroxycoumarin, 6-chloro-4-hydroxy-2h-chromen-2-one, 4-hydroxy-6-chlorocoumarin, 6-chloro-4-hydroxy-2h-1-benzopyran-2-one, 2h-1-benzopyran-2-one,6-chloro-4-hydroxy, 6-chloro-2-hydroxychromen-4-one, 6-chloro-2-hydroxy-4h-chromen-4-one, pubchem24343, acmc-1bsui, 6-chlor-4-oxy-cumarin
PubChem CID 54686348
IUPAC Name 6-chloro-2-hydroxy-4H-chromen-4-one
SMILES OC1=CC(=O)C2=CC(Cl)=CC=C2O1

Specifications

Melting Point 263°C
Quantity 1 g
Formula Weight 196.59
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloro-4-hydroxycoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.