Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chlorohexanoic Acid 98.0+%, TCI America™
SDP

Catalog No. C150725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C150725G 25 g
C15075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C150725G Supplier TCI America Supplier No. C150725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4224-62-8
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.60
MDL Number MFCD00061103
InChI Key XWWKSLXUVZVGSP-UHFFFAOYSA-N
PubChem CID 20209
IUPAC Name 6-chlorohexanoic acid
SMILES OC(=O)CCCCCCl

Specifications

Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 150.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 6-Chlorohexanoic Acid
EINECSNumber (2)-1180
RTECSNumber MO7000000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.