Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloronicotinic Acid 98.0+%, TCI America™
SDP

Catalog No. C1116250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1116250G 250 g
C111625G 25 g
C1116100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C1116250G Supplier TCI America Supplier No. C1116250G

Chemical Identifiers

CAS 5326-23-8
Molecular Formula C6H4ClNO2
Molecular Weight (g/mol) 157.553
MDL Number MFCD00006241
InChI Key UAWMVMPAYRWUFX-UHFFFAOYSA-N
Synonym 6-chloronicotinic acid, 6-chloro-3-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 6-chloro, 6-chloro-nicotinic acid, 6-chloro nicotinic acid, 2-chloro-5-pyridinecarboxylic acid, 2-chloropyridine-5-carboxylic acid, 6-chlornicotins, 6chloronichotinicacid, 2-chloroncotinic acid
PubChem CID 79222
IUPAC Name 6-chloropyridine-3-carboxylic acid
SMILES C1=CC(=NC=C1C(=O)O)Cl

Specifications

Color White-Yellow
Quantity 250 g
Formula Weight 157.55
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloronicotinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.