Learn More
AdipoGen 6-Chloropurine (25 g)
Supplier: AdipoGen CDXC0684G025
Chemical. CAS: 87-42-3. Formula: C5H3ClN4. MW: 154.56. 6-Chloropurine is a building block in chemical synthesis and is used as an intermediate in the preparation of 9-alkylpurines via alkylation with various substituted alkyl halides in DMSO. 6-Chloropurine based compounds have been shown to display antiviral and antitumor properties.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.