Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-chloropyrimidine-2,4-diamine, 97%, Thermo Scientific™

Catalog No. BTB10268DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB10268DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTB10268DA Supplier Thermo Scientific Chemicals Supplier No. BTB10268DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 156-83-2
Molecular Formula C4H5ClN4
Molecular Weight (g/mol) 144.562
MDL Number MFCD00006097
InChI Key QJIUMVUZDYPQRT-UHFFFAOYSA-N
Synonym 4-chloro-2,6-diaminopyrimidine, 2,4-diamino-6-chloropyrimidine, 2,6-diamino-4-chloropyrimidine, 6-chloro-2,4-diaminopyrimidine, 6-chloro-pyrimidine-2,4-diamine, 2,4-pyrimidinediamine, 6-chloro, 4-chloro-2,6-diamino pyrimidine, 2-amino-6-chloro-4-pyrimidinylamine, caap, minoxidil impurity b
PubChem CID 67432
IUPAC Name 6-chloropyrimidine-2,4-diamine
SMILES C1=C(N=C(N=C1Cl)N)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
Formula Weight 144.56
Percent Purity 97%
Chemical Name or Material 6-chloropyrimidine-2, 4-diamine
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.