Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chloropyrimidine-4,5-diamine 98.0+%, TCI America™
SDP

Catalog No. C34331G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C34331G Supplier TCI America Supplier No. C34331G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4316-98-7
Molecular Formula C4H5ClN4
Molecular Weight (g/mol) 144.56
MDL Number MFCD00023270
InChI Key VNSFICAUILKARD-UHFFFAOYSA-N
Synonym 4,5-Diamino-6-chloropyrimidine
PubChem CID 78010
IUPAC Name 6-chloropyrimidine-4,5-diamine
SMILES NC1=NC=NC(Cl)=C1N

Specifications

Melting Point 259°C
Quantity 1 g
Formula Weight 144.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Chloropyrimidine-4,5-diamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.