Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Chlorouracil, 98+%, Thermo Scientific™

Catalog No. AAL0187506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0187506 5 g
AAL0187503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0187506 Supplier Thermo Scientific Chemicals Supplier No. L0187506

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Chlorouracil (4-Chlorouracil; 6-Chlorouracil) is a halogenated uracil that is useful in studies of the effects of halogenation on nucleic acid base-pair stability and alkali metal ion affinity. Reaction of 6-chlorouracil with 4-(dimethylamino)pyridine, 4-methylpyridine, and pyridin-4-yl-morpholine yielded pyridinium-substituted uracils as chlorides which were converted into pyridinium uracilates by deprotonation. These heterocyclic mesomeric betaines are cross-conjugated and thus possess separate cationic (pyridinium) and anionic (uracilate) moieties. Calculations and X-ray single crystal analyses may be used to characterize these systems and to compare the salts with the betaines.

Solubility
Soluble in Ammonium Hydroxide

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4270-27-3
Molecular Formula C4H3ClN2O2
Molecular Weight (g/mol) 146.53
MDL Number MFCD00014595
InChI Key PKUFNWPSFCOSLU-UHFFFAOYSA-N
Synonym 6-chlorouracil, 6-chloropyrimidine-2,4 1h,3h-dione, 4-chlorouracil, 6-chloropyrimidine-2,4-diol, 6-chloro-2,4 1h,3h-pyrimidinedione, 6-chloro-2,4-dihydroxypyrimidine, 6-chloro-1,2,3,4-tetrahydropyrimidine-2,4-dione, 6-chloro-2,4-pyrimidinediol, 6-chloro-1,3-dihydropyrimidine-2,4-dione, 4-chloro-2,6-dihydroxypyrimidine
PubChem CID 73272
IUPAC Name 6-chloro-1H-pyrimidine-2,4-dione
SMILES ClC1=CC(=O)NC(=O)N1

Specifications

Melting Point ∽295°C (decomposition)
Quantity 5 g
Beilstein 120492
Solubility Information Soluble in Ammonium Hydroxide
Formula Weight 146.53
Percent Purity ≥98%
Chemical Name or Material 6-Chlorouracil
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 224-258-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.