Learn More
Selleck Chemical LLC 6-Diazo-5-oxo-L-norleucine-2mg
Supplier: Selleck Chemical LLC S86202MG
6-Diazo-5-oxo-L-norleucine (Diazooxonorleucine, L-6-Diazo-5-oxonorleucine, DON), an antibiotic isolated from Streptomyces, is a glutaminase antagonist with IC50 of ~1 mM for cKGA (kidney-type glutaminase). 6-Diazo-5-oxo-L-norleucine exhibits analgesic, antibacterial, antiviral and anticancer properties.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.