Learn More
AdipoGen 6-FAM
SDP
Supplier:  AdipoGen CDXC0016G001
Encompass_Preferred
Chemical. CAS 3301-79-9. Formula C21H12O7. MW 376.32. Synthetic. The amine-reactive fluorescein derivatives are probably the most common fluorescent derivatization reagents for covalently labeling proteins. Carboxyfluorescein is better retained in cells than is fluorescein, its pKa of ~6.5 is lower than the cytosolic pH of most cells pH ~6.8-7.4. Carboxyfluorescein is commonly employed as a polar tracer.
Catalog No. 50-256-1547
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.