Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Fluorochromone-2-carboxylic Acid 97.0+%, TCI America™
SDP

Catalog No. F074425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F074425G 25 g
F07445G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F074425G Supplier TCI America Supplier No. F074425G

Chemical Identifiers

CAS 99199-59-4
Molecular Formula C10H5FO4
Molecular Weight (g/mol) 208.144
MDL Number MFCD03070543
InChI Key JZJYDFADRMBXAW-UHFFFAOYSA-N
Synonym 6-fluorochromone-2-carboxylic acid, 6-fluoro-4-oxo-4h-chromene-2-carboxylic acid, 6-fluoro-4-oxo-4h-1-benzopyran-2-carboxylic acid, 6-fluoro-4-oxo-1-benzopyran-2-carboxylic acid, 6-fluoro-4-oxo-chromene-2-carboxylic acid, 4h-1-benzopyran-2-carboxylic acid, 6-fluoro-4-oxo, 6-fluorochromone-2-carboxylicacid, pubchem9710, acmc-209sbz
PubChem CID 4096046
IUPAC Name 6-fluoro-4-oxochromene-2-carboxylic acid
SMILES C1=CC2=C(C=C1F)C(=O)C=C(O2)C(=O)O

Specifications

Melting Point 258°C
Color White-Yellow
Quantity 25 g
Formula Weight 208.14
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Fluorochromone-2-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.