Learn More
Labconco™ 6 ft. Cell Logic™ Class II, Type B2 Biosafety Cabinet, US Model

Cell Logics are designed to meet the specific needs of cell research. Models feature a modified sash to accommodate a microscope and/or a temperature-controlled work area on the work surface to maintain cell viability. All models ensure the highest standard of safety and performance.
Supplier: Labconco™ 333681012
Description
Performance Features:
- Safe Class II airflows (100% exhaust, remote blower required) that provide personnel, product, and environmental protection.
- Cabinet interior provides ISO Class 5 (Grade A) or better working conditions.
- Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe downflow – even as HEPA filters load.
- Optimized airflow minimizes energy use and noise output to industry-leading levels.
- Logic OS with 5 in. line-of-sight touchscreen display.
- Access cabinet information such as filter life, alerts and alarms, data logs.
- Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet.
- Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work.
- Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work.
- Remote Communication Board for direct communication with remote blower and/or building management system featuring dry contact relays, USB data output and RS-232 RJ45 data output.
- Accessory Package: UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals.
- Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach.
- Full, six-year warranty on all Class II biosafety cabinets.
Optional Configurations:
- Scope-Ready™ Package consisting of 7.0 in. wide x 10.5 in. high (18 x 27 cm) sash cutout and Pure-Vu™ Seal with powder-coated steel collar.
- Stand-Still™ Isolation Platform is required and sold separately.
- Temp-Zone™ Package consisting of temperature-controlled work area, 10.5 in. wide x 12.0 in. deep (27 x 30 cm) located on the left side of the work surface.
- Water circulator is required (not included).
- Telescoping base stand with shelf and leveling feet.
Specifications
6 ft. Cell Logic Class II, Type B2 Biosafety Cabinet | |
Stainless Steel (304) | |
Class II, Type B2 Bio Safety Cabinet | |
67 dBA | |
North America (230 V) | |
2 Double, Left and Right Side | |
20 cm | |
2348 m³/hr. | |
99.99% | |
199.1 cm | |
LED, UV | |
8 in. | |
1382 CFM | |
72.5 in. | |
25.7 in. | |
27 in. | |
57.2 cm | |
32 in. | |
230 V |
ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2 | |
184.2 cm | |
Cell Logic | |
2 Duplex | |
360 W | |
72.7 in | |
78.4 in. | |
5 in. touchscreen | |
HEPA | |
Cabinet, Accessory Package, Scope-Ready | |
184.7 cm | |
1 | |
6 Years | |
50/60 Hz | |
22.5 in. | |
65.3 cm | |
69 cm | |
81.3 cm | |
360 W |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.