Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ 6 ft. Cell Logic™ Class II, Type B2 Biosafety Cabinet, US Model
SDP

Catalog No. 01812509 Shop All Labconco Corporation Products
Change view
Click to view available options
Includes:
Cabinet, Accessory Package, Scope-Ready
Cabinet, Accessory Package, Scope-Ready, Base Stand
Cabinet, Accessory Package, Scope-Ready, Temp-Zone
Cabinet, Accessory Package, Scope-Ready, Temp-Zone, Base Stand
Cabinet, Accessory Package, Temp-Zone
Cabinet, Accessory Package, Temp-Zone, Base Stand
Plug Type:
North America (115 V, 15 A)
North America (230 V)
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Plug Type
01-812-509 Cabinet, Accessory Package, Temp-Zone North America (115 V, 15 A)
01-812-471 Cabinet, Accessory Package, Scope-Ready North America (115 V, 15 A)
01-812-469 Cabinet, Accessory Package, Scope-Ready, Base Stand North America (115 V, 15 A)
01-812-447 Cabinet, Accessory Package, Scope-Ready, Temp-Zone North America (115 V, 15 A)
01-812-445 Cabinet, Accessory Package, Scope-Ready, Temp-Zone, Base Stand North America (115 V, 15 A)
01-812-507 Cabinet, Accessory Package, Temp-Zone, Base Stand North America (115 V, 15 A)
01-812-470 Cabinet, Accessory Package, Scope-Ready North America (230 V)
01-812-468 Cabinet, Accessory Package, Scope-Ready, Base Stand North America (230 V)
01-812-446 Cabinet, Accessory Package, Scope-Ready, Temp-Zone North America (230 V)
01-812-444 Cabinet, Accessory Package, Scope-Ready, Temp-Zone, Base Stand North America (230 V)
01-812-508 Cabinet, Accessory Package, Temp-Zone North America (230 V)
01-812-506 Cabinet, Accessory Package, Temp-Zone, Base Stand North America (230 V)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 01-812-509 Supplier Labconco™ Supplier No. 323681002
Only null left
Add to Cart
Add to Cart
Request a Quote

Cell Logics are designed to meet the specific needs of cell research. Models feature a modified sash to accommodate a microscope and/or a temperature-controlled work area on the work surface to maintain cell viability. All models ensure the highest standard of safety and performance.

Performance Features:

  • Safe Class II airflows (100% exhaust, remote blower required) that provide personnel, product, and environmental protection.
    • Cabinet interior provides ISO Class 5 (Grade A) or better working conditions.
  • Constant Airflow Profile™ (CAP) and BalanceFlow™ technology maintain safe downflow – even as HEPA filters load.
    • Optimized airflow minimizes energy use and noise output to industry-leading levels.
  • Logic OS with 5 in. line-of-sight touchscreen display.
    • Access cabinet information such as filter life, alerts and alarms, data logs.
    • Remote connectivity, particle counter integration, and certification records elevate user control of the cabinet.
  • Smart-Start™ and Night-Smart™ modes simplify startup and shut-down procedures to help ensure consistency and cleanliness before, during, and after work.
  • Inclination Technology™ improves ergonomic working conditions by maximizing comfort and accessibility during long periods of work.
  • Remote Communication Board for direct communication with remote blower and/or building management system featuring dry contact relays, USB data output and RS-232 RJ45 data output.
  • Accessory Package: UV lamp (254 nm), right-side service fixture with ball-type valve and right and left-side Vacu-Pass™ Cord and Cable Portals.
  • Mag-Zone™ area and mating accessories keep common items such as pipettes and biohazard waste organized and within reach.
  • Full, six-year warranty on all Class II biosafety cabinets.

Optional Configurations:

  • Scope-Ready™ Package consisting of 7.0 in. wide x 10.5 in. high (18 x 27 cm) sash cutout and Pure-Vu™ Seal with powder-coated steel collar.
    • Stand-Still™ Isolation Platform is required and sold separately.
  • Temp-Zone™ Package consisting of temperature-controlled work area, 10.5 in. wide x 12.0 in. deep (27 x 30 cm) located on the left side of the work surface.
    • Water circulator is required (not included).
  • Telescoping base stand with shelf and leveling feet.

Specifications

Description 6 ft. Cell Logic Class II, Type B2 Biosafety Cabinet
Certifications/Compliance ETL, UL, CAN/CSA, ADA, ISO 5 Conditions per ISO 14644-1 and 2
Chamber Material Stainless Steel (304)
Width (Metric) Interior 184.2 cm
Product Type Class II, Type B2 Bio Safety Cabinet
Model Cell Logic
Noise Level 67 dBA
Outlets 2 Duplex
Plug Type North America (115 V, 15 A)
Power Consumption 360 W
Receptacles 2 Double, Left and Right Side
Height (English) 72.7 in
Sash Opening (Metric) 20 cm
Width (English) Exterior 78.4 in.
Volume (Metric) Exhaust 2348 m³/hr.
Display 5 in. touchscreen
Filter Efficiency 99.99%
Filter Type HEPA
Width (Metric) Exterior 199.1 cm
Includes Cabinet, Accessory Package, Temp-Zone
Lighting UV, LED
Height (Metric) 184.7 cm
Sash Opening (English) 8 in.
Service Fixtures 1
Volume 1382 CFM
Warranty 6 Years
Width (English) Interior 72.5 in.
Frequency 60 Hz
Height (English) Interior 25.7 in.
Height (English) Max. Opening 22.5 in.
Height (English) Viewing 27 in.
Height (Metric) Interior 65.3 cm
Height (Metric) Max. Opening 57.2 cm
Height (Metric) Viewing 69 cm
Depth (English) 32 in.
Depth (Metric) 81.3 cm
Voltage 115 V
Energy Consumption (Operating Set Point) 360 W
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.