Learn More
Labconco 6 ft. Nexus Horizontal Clean Bench



Nexus Horizontal Clean Benches are designed to protect non-hazardous applications by providing an ISO Class 4 clean airflow environment. The large, 5 in. touchscreen interface features Nexus OS™ software that simplifies operation, maintains records and provides user customizaton of the system.
Supplier: Labconco 5360210
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Description
- 6 ft. Nexus Horizontal Clean Bench
- ISO Class 4 laminar airflow is 10x cleaner than the industry standard ISO Class 5
- Energy efficient ECM motor with CAP™ Technology ensures accurate airflow between certifications
Specifications
Horizontal Clean Bench | |
HEPA | |
Epoxy-coated Steel | |
21 in. | |
50.1 in. | |
127.5 cm | |
UV Lamp | |
NEMA 5-20P | |
75.2 in. | |
191.1 cm |
Epoxy | |
UV | |
Optic White | |
53.8 cm | |
30.2 in. | |
76.7 cm | |
None | |
230 V | |
73.0 in | |
185.4 cm |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.