Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ 6 ft. Prism Class II, Type A2 Biosafety Cabinet with 10 in. Sash and Base Stand
SDP

Catalog No. 10369236 Shop All Labconco Corporation Products
Change view
Click to view available options
Plug Type:
China, Australia (230 V)
India (230 V)
Japan (100 V)
North America (115 V, 20 A)
North America (230 V)
Schuko (230 V)
UK (230 V)
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Plug Type
10-369-236 North America (230 V)
10-369-220 North America (115 V, 20 A)
10-369-222 Schuko (230 V)
10-369-221 Japan (100 V)
10-369-211 India (230 V)
10-369-210 UK (230 V)
10-369-209 China, Australia (230 V)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 10-369-236 Supplier Labconco™ Supplier No. 302617112
Only null left
Add to Cart
Add to Cart
Request a Quote

Prism™ Class II, Type A2 Biosafety Cabinet delivers protection where it matters most - for you, your work, and your lab environment. Engineered for performance, designed for usability, and built to last, Prism BSCs make high-level biosafety accessible without compromise.

  • Prism offers complete personnel, product, and environmental protection
  • NSF/ANSI 49 listed for regulatory compliance
  • Constant Airflow Profile™ (CAP) airflow monitoring system maintains airflow setpoints in real time as filters load
  • BalanceFlow™ plenum provides energy-efficiency and ultra-quiet operation
  • Prism OS with 5 in. line-of-sight touchscreen display. Access cabinet information such as filter life, alerts and alarms, data logs. Remote connectivity and certification records elevate user control of the cabinet
  • Containment of biohazards in BSL 1-4 environments

Specifications

Certifications/Compliance NSF/ANSI 49, UL 61010-1, CAN/CSA C22.2, CE Marked, ADA
Chamber Material Stainless Steel (304)
Product Type Class II Biosafety Cabinet
Model Prism Class II
Noise Level 64 dBA
Outlets 2 Duplex
Plug Type North America (230 V)
Power Consumption 418 W
Receptacles 2 Duplex, GFI
Sash Opening (Metric) 20 cm
Volume (Metric) Exhaust 1308 m3/hr.
Frequency 50/60 Hz
Voltage 115 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.