Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ 6 ft. Purifier Axiom Class II Type C1 Biosafety Cabinet with 10 in. Sash Opening
SDP

6 ft. Purifier Axiom Class II Type C1 Biosafety Cabinet with 10 in. Sash Opening is an unique, flexible BSC that can be operated as Type A or B and is the only Type C1 in the world.

$24430.00 - $26400.00

Specifications

Description Class II Type C1 Biosafety Cabinet
Certifications/Compliance NSF/ANSI 49, UL 61010, CAN/CSA C22.2, CE Marked, ISO 5 Conditions per ISO 14644-1 and 2, ADA
Chamber Material Stainless Steel
Product Type Class II Biosafety Cabinet
Model Axiom
View More Specs

Products 20
Catalog Number Mfr. No. Plug Type Electrical Requirements Includes Price Quantity & Availability  
Catalog Number Mfr. No. Plug Type Electrical Requirements Includes Price Quantity & Availability  
30-461-0021
SDP
View Documents
Labconco™
304610021
Japan 100 V, 50/60 Hz, 12 A Biosafety Cabinet Only
Each for $24,430.00
Only null left
Add to Cart
 
30-461-0031
SDP
View Documents
Labconco™
304610031
Schuko 230 V, 50/60 Hz, 6 A Biosafety Cabinet Only
Each for $24,430.00
Only null left
Add to Cart
 
30-461-0041
SDP
View Documents
Labconco™
304610041
China, Australia 230 V, 50/60 Hz, 6 A Biosafety Cabinet Only
Each for $24,430.00
Only null left
Add to Cart
 
30-461-0051
SDP
View Documents
Labconco™
304610051
UK 230 V, 50/60 Hz, 6 A Biosafety Cabinet Only
Each for $24,430.00
Only null left
Add to Cart
 
30-461-0071
SDP
View Documents
Labconco™
304610071
India 230 V, 50/60 Hz, 6 A Biosafety Cabinet Only
Each for $24,430.00
Only null left
Add to Cart
 
30-461-0121
SDP
View Documents
Labconco™
304610121
Japan 100 V, 50/60 Hz, 12 A Base Stand
Each for $25,500.00
Only null left
Add to Cart
 
30-461-0131
SDP
View Documents
Labconco™
304610131
Schuko 230 V, 50/60 Hz, 6 A Base Stand
Each for $25,500.00
Only null left
Add to Cart
 
30-461-0141
SDP
View Documents
Labconco™
304610141
China, Australia 230 V, 50/60 Hz, 6 A Base Stand
Each for $25,500.00
Only null left
Add to Cart
 
30-461-0151
SDP
View Documents
Labconco™
304610151
UK 230 V, 50/60 Hz, 6 A Base Stand
Each for $25,500.00
Only null left
Add to Cart
 
30-461-0171
SDP
View Documents
Labconco™
304610171
India 230 V, 50/60 Hz, 6 A Base Stand
Each for $25,500.00
Only null left
Add to Cart
 
30-461-1021
SDP
View Documents
Labconco™
304611021
Japan 100 V, 50/60 Hz, 12 A Accessory Pk.
Each for $25,450.00
Only null left
Add to Cart
 
30-461-1031
SDP
View Documents
Labconco™
304611031
Schuko 230 V, 50/60 Hz, 6 A Accessory Pk.
Each for $25,450.00
Only null left
Add to Cart
 
30-461-1041
SDP
View Documents
Labconco™
304611041
China, Australia 230 V, 50/60 Hz, 6 A Accessory Pk.
Each for $25,450.00
Only null left
Add to Cart
 
30-461-1051
SDP
View Documents
Labconco™
304611051
UK 230 V, 50/60 Hz, 6 A Accessory Pk.
Each for $25,450.00
Only null left
Add to Cart
 
30-461-1071
SDP
View Documents
Labconco™
304611071
India 230 V, 50/60 Hz, 6 A Accessory Pk.
Each for $25,450.00
Only null left
Add to Cart
 
30-461-1121
SDP
View Documents
Labconco™
304611121
Japan 100 V, 50/60 Hz, 12 A Base Stand, Accessory Pk.
Each for $26,400.00
Only null left
Add to Cart
 
30-461-1131
SDP
View Documents
Labconco™
304611131
Schuko 230 V, 50/60 Hz, 6 A Base Stand, Accessory Pk.
Each for $26,400.00
Only null left
Add to Cart
 
30-461-1141
SDP
View Documents
Labconco™
304611141
China, Australia 230 V, 50/60 Hz, 6 A Base Stand, Accessory Pk.
Each for $26,400.00
Only null left
Add to Cart
 
30-461-1151
SDP
View Documents
Labconco™
304611151
UK 230 V, 50/60 Hz, 6 A Base Stand, Accessory Pk.
Each for $26,400.00
Only null left
Add to Cart
 
30-461-1171
SDP
View Documents
Labconco™
304611171
India 230 V, 50/60 Hz, 6 A Base Stand, Accessory Pk.
Each for $26,400.00
Only null left
Add to Cart
 
Description

Description

  • Requires supporting base if included stand option is not selected
  • Intrinsically-safe negative pressure design includes:
    • Dual Electronically Commutated Motors (ECM)
    • Constant Airflow Profile™ (CAP) Technology airflow monitoring system
    • Active Protection Protocol operates the cabinet's exhaust blowers for for up to five minutes preventing the flow of air into the laboratory in the event of exhaust system failure
    • BSC required Air-Wave™ Entry System at access opening to enhance containment
    • Contain-Air™ Negative Pressure Channel at top of sash to enhance containment
    • Two programmable working sash heights (8 in. and 10 in.)
    • 10° angled, fully-closing, 1/4 in. tempered safety glass sash with counterbalanced, anti-racking, easy-to-lift mechanism
    • Curved waterfall design stainless steel inlet grille with Reserve-Air™ Secondary Airflow Slots
    • 22.6 in. (58 cm) maximum sash opening height and 27.0 in. (69 cm) viewing height Chem-Zone™ work surface with dedicated direct exhaust for use with hazardous vapors or radionuclides
    • Leak-tight interior with removable, type 304 stainless steel 3-piece work surface (dished Chem-Zone in center and two flat sides) with lift out knobs and clearly delineated working area
    • Nominal inflow velocity of 105 feet per minute (fpm) (0.53 m/sec)
    • Nominal downflow velocity of 65 fpm (0.33 m/sec) for 4 ft. models and 55 fpm (0.28 m/sec) for 6 ft. models
  • Touchpad on right-hand side post controls blower, light(s), timer, electrical receptacles, audible alarm mute and menu selection
  • Omni-Flex™ two mode design (Type A and B mode)
    • Low static pressure and volumetric rate exhaust requirements when in Type B mode
    • Night-Smart™ System for idling the blower when the sash is fully closed (while in Type A mode only)
  • MyLogic™ Operating System and interior-mounted, line-of-sight, full color, easy-to-understand LCD information center with:
    • Smart-Start™ System for start up and shut down operations
    • Filter Life Remaining bar graph for each HEPA filter
    • Status line for alarm conditions and alerts to warn when filter life diminishes to 20% and 0%
    • Bright, 90-150 ft. candle, glare-free LED lighting located outside the contaminated work area
    • Interval/elapsed timer for experiment monitoring, LED light or UV light control (on models with UV light)
    • Electronic security lock (optional activation) that requires access code to operate the cabinet
Specifications

Specifications

Class II Type C1 Biosafety Cabinet
Stainless Steel
Axiom
2
2
78.3 in.
Full-color LCD
HEPA
Powder Coated Steel
10 in.
64.2 in.
32.7 in.
955 lb.
NSF/ANSI 49, UL 61010, CAN/CSA C22.2, CE Marked, ISO 5 Conditions per ISO 14644-1 and 2, ADA
Class II Biosafety Cabinet
67 dBA
480 W
25 cm
968 m3/hr.
0.9999
198.9 cm
LED
5 Years
163.1 cm
83.1 cm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.