Learn More
Cayman Chemical 6-Gingerol, 23513-14-6, 1 mg
SDP
Supplier:  Cayman Chemical 117071
Encompass_Preferred
Molecular Formula C17H26O4, Purity ≥98%, Formula Weight 294.4, Synonyms: 5S-hydroxy-1-(4-hydroxy-3-methoxyphenyl)-3-decanone; A natural chemical found in fresh ginger that activates TRPV1 (EC50 = 3.3 M) as well as the TRP ankyrin receptor TRPA1 (EC50 = 10.4 M).
Catalog No. NC1301318
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.