Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Hydroxy-2-naphthoic Acid 98.0+%, TCI America™
SDP

Catalog No. H0612500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0612500G 500 g
H06125G 5 g
H061225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0612500G Supplier TCI America Supplier No. H0612500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16712-64-4
Molecular Formula C11H8O3
Molecular Weight (g/mol) 188.182
MDL Number MFCD00060070
InChI Key KAUQJMHLAFIZDU-UHFFFAOYSA-N
Synonym 6-hydroxy-2-naphthoic acid, 2-naphthalenecarboxylic acid, 6-hydroxy, 6-hydroxy-2-naphthalenecarboxylic acid, 6-carboxy-2-naphthol, 2-hydroxynaphthalene-6-carboxylic acid, 6-hydroxy-beta-naphthoic acid, unii-72xa858i6p, 6-hydroxy-naphthalene-2-carboxylic acid, 2-naphthoic acid, 6-hydroxy, 2-hydroxy-6-naphthoic acid
PubChem CID 85557
IUPAC Name 6-hydroxynaphthalene-2-carboxylic acid
SMILES C1=CC2=C(C=CC(=C2)O)C=C1C(=O)O

Specifications

Melting Point 248°C
Quantity 500 g
Formula Weight 188.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Hydroxy-2-naphthoic Acid
EINECSNumber (4)-0398
RTECSNumber QN3722500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.