Learn More
Medchemexpress LLC HY-W011956 5mg Medchemexpress, 6-Hydroxymelatonin CAS:2208-41-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW0119565MG
Encompass_Preferred
Medchemexpress, HY-W011956 5mg 6-Hydroxymelatonin CAS:2208-41-5 6-Hydroxymelatonin is a primary metabolic of Melatonin, which is metabolized by cytochrome P450 (CYP) 1A2. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3320
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.