Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam 6-IAF, Thiol-reactive Fluorescent Label, Solid, 50 mg

Catalog No. 502705173
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5173 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5173 Supplier Abcam Supplier No. AB14546450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Thiol-reactive fluorescent label for FRET studies, actin, and protein labeling.

  • Molecular Weight: 515.3 Da
  • Purity: >95%
  • Form: Solid
  • Properties: Thiol-reactive fluorescent label
  • Application: Acts as an acceptor fluorophore in FRET studies
  • Excitation and Emission Maxima: 347 and 530 nm
  • Derivative: 6-IATR (ab145010) derivative
  • Labeling Targets: Actin, microtubule-associated proteins and histones
  • Storage Temperature: Store at Ambient

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Actin, Microtubule-associated Proteins, Histones
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Form Solid
Label or Dye Fluorescein
Product Type Protein Labeling Reagent
Buffer 100% N-(3',6'-dihydroxy-1-oxospiro[2-benzofuran-3,9'-xanthene]-5-yl)-2-iodoacetamide
Content And Storage Store at ambient temperature.
Detection Method Fluorescence
Formulation 100% N-(3',6'-dihydroxy-1-oxospiro[2-benzofuran-3,9'-xanthene]-5-yl)-2-iodoacetamide
Quantity 50 mg
For Use With (Application) FRET studies, Labeling actin, Labeling microtubule-associated proteins, Labeling histones
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.