Learn More
Abcam 6-Iodoacetamidofluorescein, Thiol-Reactive Fluorescent Label, Solid, >95% Purity, 250 mg
Description
- Molecular Weight: 515.3 Da
- Purity: >95%
- Physical Form: Solid
- Chemical Properties: Thiol-reactive fluorescent label
- Applications: Acts as an acceptor fluorophore in FRET studies
- Optical Properties: Excitation and emission maxima of 347 and 530 nm
- Target Specificity: Labels actin, microtubule-associated proteins and histones
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Thiols |
| Shipping Condition | Ambient |
| Substrate Properties | Chemical Substrate |
| Label or Dye | Fluorescein |
| Buffer | 100% N-(3',6'-dihydroxy-1-oxospiro[2-benzofuran-3,9'-xanthene]-5-yl)-2-iodoacetamide |
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Detection Method | Fluorescence |
| Quantity | 250 mg |
| For Use With (Application) | FRET studies, Labeling actin, Labeling microtubule-associated proteins, Labeling histones |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.