Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Maleimidohexanehydrazide Trifluoroacetate 95.0+%, TCI America™
SDP

Catalog No. M273550MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M273550MG 50 mg
M2735250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M273550MG Supplier TCI America Supplier No. M273550MG

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 151038-94-7
Molecular Formula C12H16F3N3O5
Molecular Weight (g/mol) 339.27
MDL Number MFCD07357647
InChI Key DRURMGRBAVYWCL-UHFFFAOYSA-N
Synonym epsilon-Maleimidocaproyl Hydrazide Trifluoroacetate, EMCH
PubChem CID 23509306
IUPAC Name 6-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)hexanehydrazide; trifluoroacetic acid
SMILES OC(=O)C(F)(F)F.NNC(=O)CCCCCN1C(=O)C=CC1=O

Specifications

Color White-Yellow
Quantity 50 mg
Formula Weight 339.27
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Maleimidohexanehydrazide Trifluoroacetate
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.