Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methyl-1,2,3,4-tetrahydroquinoline 97.0+%, TCI America™
SDP

Catalog No. M130225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M130225G 25 g
M13025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M130225G Supplier TCI America Supplier No. M130225G

Chemical Identifiers

CAS 91-61-2
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.22
MDL Number MFCD00023887
InChI Key XOKMRXSMOHCNIX-UHFFFAOYSA-N
Synonym civettal, 1,2,3,4-tetrahydro-6-methylquinoline, quinoline, 1,2,3,4-tetrahydro-6-methyl, quinoline, tetrahydro-6-methyl, p-methyltetrahydroquinoline, acmc-209rck, dsstox_cid_27444, dsstox_rid_82351, dsstox_gsid_47444, xokmrxsmohcnix-uhfffaoysa
PubChem CID 66678
IUPAC Name 6-methyl-1,2,3,4-tetrahydroquinoline
SMILES CC1=CC=C2NCCCC2=C1

Specifications

Melting Point 35°C
Color Yellow
Quantity 25 g
Formula Weight 147.22
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 6-Methyl-1,2,3,4-tetrahydroquinoline
EINECSNumber (5)-0803
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.