Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methyl-1-indanone 98.0+%, TCI America™
SDP

Catalog No. M190125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M190125G 25 g
M19015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M190125G Supplier TCI America Supplier No. M190125G

Chemical Identifiers

CAS 24623-20-9
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00275729
InChI Key DBOXRDYLMJMQBB-UHFFFAOYSA-N
Synonym 6-methyl-1-indanone, 6-methyl-2,3-dihydro-1h-inden-1-one, 6-methylindan-1-one, 6-methyl-indan-1-one, 1h-inden-1-one, 2,3-dihydro-6-methyl, 2,3-dihydro-6-methylinden-1-one, pubchem8851, acmc-209gdm, 6-methyl-indane-1-one
PubChem CID 2795093
IUPAC Name 6-methyl-2,3-dihydro-1H-inden-1-one
SMILES CC1=CC2=C(CCC2=O)C=C1

Specifications

Melting Point 62°C
Color White-Yellow
Boiling Point 126°C
Quantity 25 g
Formula Weight 146.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methyl-1-indanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.