Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-methyl-2,3-dihydropyridazin-3-one hydrate, Thermo Scientific™

Catalog No. p-4391400
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB06840DA 1 g
BTB06840EA 10 g
BTB06840EE 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BTB06840DA Supplier Thermo Scientific Chemicals Supplier No. BTB06840DA
Only null left
Add to Cart

Chemical Identifiers

CAS 7143-82-0
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD00039720
InChI Key QZWIXLPWMGHDDD-UHFFFAOYSA-N
Synonym 6-methylpyridazin-3-ol hydrate, 6-methylpyridazin-3 2h-one hydrate, 6-methyl-2,3-dihydropyridazin-3-one hydrate, 6-methyl-2h-pyridazin-3-one hydrate, 6-methylpyridazin-3-one hydrate, 3-methyl-1h-pyridazin-6-one hydrate, 6-methylpyridazin-3 2h ;-one hydrate, 6-methyl-2-hydropyridazin-3-one, hydrate, 6-methyl-2,3-dihydropyridazine-3-one hydrate, 6-methylpyridazin-3 2h-one-water 1/1
PubChem CID 2774632
SMILES CC1=NNC(=O)C=C1

Specifications

Assay Percent Range 90%
Quantity 1 g
IUPAC Name 6-methyl-2,3-dihydropyridazin-3-one
Formula Weight 128.14
Percent Purity 90%
Chemical Name or Material 6-Methyl-2, 3-dihydropyridazin-3-one hydrate
Hazard Category Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.