Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methyl-2-(p-tolylamino)-4H-3,1-benzoxazin-4-one 95.0+%, TCI America™
SDP

Catalog No. M257210MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M257210MG 10 mg
M257250MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M257210MG Supplier TCI America Supplier No. M257210MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 86672-58-4
Molecular Formula C16H14N2O2
Molecular Weight (g/mol) 266.30
MDL Number MFCD03272508
InChI Key GFWNGVKCDGYFKG-UHFFFAOYSA-N
Synonym URB-754, 6-Methyl-2-(p-tolylamino)-4H-benzo[d][1,3]oxazin-4-one
PubChem CID 848487
IUPAC Name 6-methyl-2-[(4-methylphenyl)amino]-4H-3,1-benzoxazin-4-one
SMILES CC1=CC=C(NC2=NC3=CC=C(C)C=C3C(=O)O2)C=C1

Specifications

Melting Point 215°C
Color Yellow
Quantity 10 mg
Formula Weight 266.30
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methyl-2-(p-tolylamino)-4H-3,1-benzoxazin-4-one
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.