Learn More
Sigma Organic Chemistry 6-Methyl-3-heptanol | 250mg | 18720-66-6 | MFCD00046682
Supplier: Sigma Organic Chemistry CDS000278250MG

6-Methyl-3-heptanol , 250mg
About this Item:
CAS #: 18720-66-6
MDL #: MFCD00046682
Molecular Weight: 130.23
UNSPSC Code: 12352200
Chemical Formula: C8H18O
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.