Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methylchromone-3-carbonitrile 98.0+%, TCI America™
SDP

Catalog No. M24591G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24591G 1 g
M24595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24591G Supplier TCI America Supplier No. M24591G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 50743-18-5
Molecular Formula C11H7NO2
Molecular Weight (g/mol) 185.18
MDL Number MFCD00191962
InChI Key FNRLRLMPBIRUSQ-UHFFFAOYSA-N
Synonym 3-cyano-6-methylchromone, 6-methylchromone-3-carbonitrile, 6-methyl-4-oxo-4h-1-benzopyran-3-carbonitrile, 4h-1-benzopyran-3-carbonitrile, 6-methyl-4-oxo, 6-methyl-4-oxo-4h-chromene-3-carbonitrile, acmc-1at8c, 6-methylchromanone-3-carbonitrile, 6-methyl-4-oxo-1-benzopyran-3-carbonitrile, 4-oxo-6-methyl-4h-1-benzopyran-3-carbonitrile
PubChem CID 688705
IUPAC Name 6-methyl-4-oxo-4H-chromene-3-carbonitrile
SMILES CC1=CC=C2OC=C(C#N)C(=O)C2=C1

Specifications

Melting Point 152°C
Color Yellow
Quantity 1 g
UN Number 3439
Formula Weight 185.18
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methylchromone-3-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.