Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methylflavone 98.0+%, TCI America™
SDP

Catalog No. M27661G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27661G 1 g
M27665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27661G Supplier TCI America Supplier No. M27661G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 29976-75-8
Molecular Formula C16H12O2
Molecular Weight (g/mol) 236.27
MDL Number MFCD00017461
InChI Key NOQJBXPAMJLUSS-UHFFFAOYSA-N
PubChem CID 689013
IUPAC Name 6-methyl-2-phenylchromen-4-one
SMILES CC1=CC2=C(C=C1)OC(=CC2=O)C3=CC=CC=C3

Specifications

Melting Point 121°C
Color White-Yellow
Boiling Point 235°C
Quantity 1 g
Formula Weight 236.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methylflavone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.