Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methylindole, 98%

Catalog No. AAL1750903
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1750903 1 g
AAL17509MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1750903 Supplier Thermo Scientific Chemicals Supplier No. L1750903
Add to Cart

CAS: 3420-02-8 | C9H9N | 131.18 g/mol

It is a reactant for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators, reactant for preparation of indole linked triazole derivatives as antifungal agents, reactant for preparation of N-β-D-xylosyl-indole derivatives as SGLT2 inhibitors for management of hyperglycemia in diabetes, reactant for preparation of aminoguanidine derivatives of arylsulfonylacylindoles as antifungal agents, reactant for preparation of indolylindazoles and indolylpyrazolopyridines as interleukin-2 inducible T cell kinase inhibitors, reactant for preparation of arylsulfonylacetylindoles as anti-HIV-1 agents. 6-Methylindole was used in the synthesis of benz[c,d]indol-3(1H)-one derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reactant for preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators, reactant for preparation of indole linked triazole derivatives as antifungal agents, reactant for preparation of N-β-D-xylosyl-indole derivatives as SGLT2 inhibitors for management of hyperglycemia in diabetes, reactant for preparation of aminoguanidine derivatives of arylsulfonylacylindoles as antifungal agents, reactant for preparation of indolylindazoles and indolylpyrazolopyridines as interleukin-2 inducible T cell kinase inhibitors, reactant for preparation of arylsulfonylacetylindoles as anti-HIV-1 agents. 6-Methylindole was used in the synthesis of benz[c,d]indol-3(1H)-one derivatives.

Solubility
Insoluble in water.

Notes
Moisture and light sensitive. Store in dry condition. Keep away from active metals, bases, oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3420-02-8
MDL Number MFCD00005682

Specifications

Melting Point 29°C to 32°C
Density 1.059
Boiling Point 112°C (5 mmHg)
Flash Point >110°C (230°F)
Molecular Formula C9H9N
Refractive Index 1.6071
Quantity 1 g
Beilstein 109708
Sensitivity Light sensitive
Synonym 6-methylindole, 1h-indole, 6-methyl, 6-methyl indole, indole, 6-methyl, pubchem7240, 6-methyl-indole, acmc-1cokr, #, ksc491i2r
Solubility Information Insoluble in water.
InChI Key ONYNOPPOVKYGRS-UHFFFAOYSA-N
SMILES CC1=CC2=C(C=C1)C=CN2
IUPAC Name 6-methyl-1H-indole
Molecular Weight (g/mol) 131.18
PubChem CID 137928
Formula Weight 131.18
Percent Purity 98%
Chemical Name or Material 6-Methylindole
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.