Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Methylnicotinic Acid 98.0+%, TCI America™
SDP

Catalog No. M189425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M189425G 25 g
M18945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M189425G Supplier TCI America Supplier No. M189425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3222-47-7
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00006341
InChI Key RZOKQIPOABEQAM-UHFFFAOYSA-N
Synonym 6-methylnicotinic acid, 3-pyridinecarboxylic acid, 6-methyl, 6-methyl nicotinic acid, 2-methylpyridine-5-carboxylic acid, 2-methyl-5-pyridinecarboxylic acid, 6-methyl-3-pyridinecarboxylic acid, 2-methyl-5-pyridine carboxylic acid, 6-methylnicotinate, 6-methylnicotinicacid, pubchem1218
PubChem CID 137860
IUPAC Name 6-methylpyridine-3-carboxylic acid
SMILES CC1=NC=C(C=C1)C(=O)O

Specifications

Melting Point 210°C
Quantity 25 g
Formula Weight 137.14
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Methylnicotinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.