Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BTX™ 6-needle Array Electrodes for BTX™ AgilePulse™ In Vivo

Catalog No. BTX470060 Shop All BTX Harvard Products
Change view
Click to view available options
Size (Metric) Gap:
4 mm
6 mm
Length (Metric):
2 mm
10 mm
16 mm
25 mm
Tip Type:
Fine Point
Trocar Point
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric) Gap Length (Metric) Tip Type
BTX470060 6 mm 2 mm Fine Point
BTX470050 4 mm 2 mm Fine Point
BTX470070 6 mm 10 mm Fine Point
BTX470086 6 mm 16 mm Trocar Point
BTX470080 6 mm 25 mm Trocar Point
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. BTX470060 Supplier BTX™ Supplier No. 470060
Add to Cart
Add to Cart

6-needle Array Electrodes for BTX™ AgilePulse™ In Vivo are miniature needles designed to provide highly-uniform electric fields in dermal or muscular tissue as part of the AgilePulse In Vivo Gene Delivery Electroporation System. Specially designed for gene and vaccine delivery.

The miniature parallel-needle array is inserted directly into the target site for fast, reliable in vivo electroporation. The electric fields produced are the closest approximation of parallel plate electrodes, treating approximately 80% of the target area with 95% of the applied electric field.

The needle array requires the needle array handle which includes the electrode connector cable. Several needle lengths and row spacing are available for various intra-dermal or intra-muscular applications. Needle Arrays can be used for up to 500 pulses with proper care and maintenance.

  • Uniform, reliable electric fields
  • Miniature needles minimize tissue trauma
  • Disposable and sterile
  • Medical-grade plastic and surgical steel construction
  • Safety-assured design
  • Multiple configurations, sizes to choose from

Specifications

For Use With (Application) Gene and Vaccine Delivery
For Use With (Equipment) AgilePulse™ in Vivo Gene Delivery Electroporation System
Material Stainless Steel
Product Line BTX Electroporation
Product Type Electroporation Needle Array
Description 6-needle Array, 6 mm gap, 2 mm length
Diameter (English) 0.01 in.
Diameter (Metric) 0.3 mm
Electrode Shape Straight/Needle Array
Includes 6-needle Array
Length (English) 0.08 in.
Length (Metric) 2 mm
Size (Metric) Gap 6 mm
Tip Type Fine Point
Sterility Sterile
Connection Type Electrode Connector Cable
Pulse Form Square Wave
Show More Show Less

Intended for Research use only. Not for use in diagnostic or clinical procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.