Learn More
Description
The miniature parallel-needle array is inserted directly into the target site for fast, reliable in vivo electroporation. The electric fields produced are the closest approximation of parallel plate electrodes, treating approximately 80% of the target area with 95% of the applied electric field.
The needle array requires the needle array handle which includes the electrode connector cable. Several needle lengths and row spacing are available for various intra-dermal or intra-muscular applications. Needle Arrays can be used for up to 500 pulses with proper care and maintenance.
- Uniform, reliable electric fields
- Miniature needles minimize tissue trauma
- Disposable and sterile
- Medical-grade plastic and surgical steel construction
- Safety-assured design
- Multiple configurations, sizes to choose from
Specifications
Specifications
| For Use With (Application) | Gene and Vaccine Delivery |
| For Use With (Equipment) | AgilePulse™ in Vivo Gene Delivery Electroporation System |
| Material | Stainless Steel |
| Product Line | BTX Electroporation |
| Product Type | Electroporation Needle Array |
| Description | 6-needle Array, 6 mm gap, 25 mm length |
| Diameter (English) | 0.03 in. |
| Diameter (Metric) | 0.7 mm |
| Electrode Shape | Straight/Needle Array |
| Includes | 6-needle Array |
| Show More |
Intended for Research use only. Not for use in diagnostic or clinical procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.