Learn More
Sigma Organic Chemistry 6-Nitro-2H-pyrido 3 2-b 1G
Supplier: Sigma Organic Chemistry OTV0008521G
6-Nitro-2H-pyrido 3 2-b , 1G
About this item:
MDL #: MFCD10000806
Chemical Formula: C7H5N3O4
Molecular Weight: 195.13
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.