Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Nitrocoumarin 98.0+%, TCI America™
SDP

Catalog No. N092325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N092325G 25 g
N09235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N092325G Supplier TCI America Supplier No. N092325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2725-81-7
Molecular Formula C9H5NO4
Molecular Weight (g/mol) 191.142
MDL Number MFCD00016973
InChI Key RMERXEXZXIVNBF-UHFFFAOYSA-N
Synonym 6-nitrocoumarin, 6-nitro-2h-chromen-2-one, 6-nitro-2-benzopyrone, 2h-1-benzopyran-2-one, 6-nitro, acmc-20abd6, maybridge1_002226, amtgc084, 6-nitro-2h-chromen-2-one #, 2h-1-benzopyran-2-one,6-nitro, 6-hydroxy oxido amino-2h-chromen-2-one
PubChem CID 75944
IUPAC Name 6-nitrochromen-2-one
SMILES C1=CC2=C(C=CC(=O)O2)C=C1[N+](=O)[O-]

Specifications

Melting Point 191°C
Color White-Yellow
Quantity 25 g
Formula Weight 191.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Nitrocoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.