Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Nitrofluorescein (isomer II) 98.0+%, TCI America™
SDP

Catalog No. N04451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N04451G 1 g
N04455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N04451G Supplier TCI America Supplier No. N04451G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27402-68-2
Molecular Formula C20H11NO7
Molecular Weight (g/mol) 377.308
MDL Number MFCD00059694
InChI Key GYWGQAIDMYGOLU-UHFFFAOYSA-N
PubChem CID 117391
IUPAC Name 3',6'-dihydroxy-5-nitrospiro[2-benzofuran-3,9'-xanthene]-1-one
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C3(C4=C(C=C(C=C4)O)OC5=C3C=CC(=C5)O)OC2=O

Specifications

Quantity 1 g
Synonym 6-Nitro-3′C,6′C-dihydroxyspiro[isobenzofuran-1(3H),9′C-(9H)xanthen]-3-one
Formula Weight 377.31
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Nitrofluorescein (isomer II)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.