Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Nitroquinoline 98.0+%, TCI America™
SDP

Catalog No. N025225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N025225G 25 g
N02525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N025225G Supplier TCI America Supplier No. N025225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 613-50-3
Molecular Formula C9H6N2O2
Molecular Weight (g/mol) 174.16
MDL Number MFCD00006799
InChI Key SMHPLBXIVNQFBA-UHFFFAOYSA-N
Synonym quinoline, 6-nitro, 6-nitro-quinoline, ccris 456, 6-nitro quinoline, zlchem 253, pubchem20822, acmc-1bcwl, dsstox_cid_984, 6-nitroquinoline, dsstox_rid_75906
PubChem CID 11945
IUPAC Name 6-nitroquinoline
SMILES [O-][N+](=O)C1=CC=C2N=CC=CC2=C1

Specifications

Melting Point 151°C
Quantity 25 g
Formula Weight 174.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 6-Nitroquinoline
RTECSNumber VC1900000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.