Description
- 6 pack of HEPA High efficient particulate filters
- Suitable for protection against airborne particulate (dust-mists and fumes)
- Captures 99.97% of particulates-up to 0.3 micron
Specifications
Specifications
| Filter Type | HEPA |
| For Use With (Equipment) | SALUS HC System |
| Quantity | 6/Pk. |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.