Learn More
AdipoGen 6-ROX
SDP
Supplier:  AdipoGen CDXC0084M025
Encompass_Preferred
Chemical. CAS 194785-18-7. Formula C33H30N2O5. MW 534.6. Synthetic. This pure-6 isomer is a derivative of carboxy-X-rhodamine, widely used for oligonucleotide labeling and automated DNA sequencing applications.
Catalog No. 50-256-1486
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.