Learn More
AnaSpec (6-TAMRA, SE) 5MG
Supplier:  AnaSpec AS81126
Encompass_Preferred
6-TAMRA, SE[6-Carboxytetramethylrhodamine, succinimidyl ester; 6-TAMRA, NHS ester] ; MW 527.53; (5 mg) 6-TAMRA, SE is the other purified single isomer of 5(6)-TAMRA, SE. It is predominantly used for nucleotide labeling and DNA sequencing.
Catalog No. NC0938217
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.