Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-(tetrahydropyran-4-yloxy)pyridine-2-carbaldehyde, Thermo Scientific™

Catalog No. CC58204DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC58204DA 1 g Amber Glass Bottle
CC58204CB 250 mg Amber Glass Bottle
CC58204DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC58204DA Supplier Thermo Scientific Chemicals Supplier No. CC58204DA
Add to Cart

Chemical Identifiers

CAS 898289-54-8
Molecular Formula C11H13NO3
Molecular Weight (g/mol) 207.229
InChI Key SBTGZJPHUYLRRU-UHFFFAOYSA-N
Synonym 6-tetrahydropyran-4-yloxy pyridine-2-carbaldehyde, 6-oxan-4-yloxy pyridine-2-carbaldehyde, 6-oxan-4-yl oxy pyridine-2-carbaldehyde, 6-tetrahydro-2h-pyran-4-yl oxy picolinaldehyde, 6-2h-3,4,5,6-tetrahydropyran-4-yloxy pyridine-2-carbaldehyde
PubChem CID 45594294
IUPAC Name 6-(oxan-4-yloxy)pyridine-2-carbaldehyde
SMILES C1COCCC1OC2=CC=CC(=N2)C=O

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 207.23
Percent Purity 95%
Chemical Name or Material 6-(tetrahydropyran-4-yloxy)pyridine-2-carbaldehyde
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.