Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-(Trifluoromethyl)nicotinic Acid 98.0+%, TCI America™
SDP

Catalog No. T22031G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T22031G 1 g
T22035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T22031G Supplier TCI America Supplier No. T22031G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 231291-22-8
Molecular Formula C7H3F3NO2
Molecular Weight (g/mol) 190.10
MDL Number MFCD00792430
InChI Key JNYLMODTPLSLIF-UHFFFAOYSA-M
Synonym 6-trifluoromethyl nicotinic acid, 6-trifluoromethyl pyridine-3-carboxylic acid, 6-trifluoromethylnicotinic acid, 6-trifluoromethyl-nicotinic acid, 2-trifluoromethyl-5-pyridinecarboxylic acid, 3-pyridinecarboxylic acid, 6-trifluoromethyl, 2-trifluoromethyl pyridine-5-carboxylic acid, 6-trifluoromethyl-3-pyridinecarboxylic acid
PubChem CID 2777551
IUPAC Name 6-(trifluoromethyl)pyridine-3-carboxylate
SMILES [O-]C(=O)C1=CC=C(N=C1)C(F)(F)F

Specifications

Melting Point 197°C
Color Yellow
Quantity 1 g
Formula Weight 191.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-(Trifluoromethyl)nicotinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.