Learn More
Terumo™ 60 cc Syringes
Catalog No. 22290023 Shop All Terumo Medical Corporation Products
Change view
Click to view available options
Connection Type:
Catheter
Eccentric Luer Slip
Luer Lock
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
22-290-023 Eccentric Luer Slip
22-290-022 Luer Lock
22-290-024 Catheter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-290-023 Supplier Terumo™ Supplier No. SS60ES
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
60 cc Syringes are available with various style tips to suit application.
  • Superior sharpness and smoothness
  • Ultra Sharp Needles
  • Clear Barrels
  • Bold, Precise Scale Markings
  • Smooth Plunger Action
  • Tamper Evident Peel Packs
Specifications
Material Plastic
Color Clear
Includes Syringe Only
Latex Free Yes
Product Type Syringe
Disposable Single-use
Graduations 1 cc
Quantity 25 Pk., 4 Pk./Cs.
Volume (Metric) 60 mL
Connection Type Eccentric Luer Slip
Graduated Yes
Sterility Sterile