Learn More
Hamilton™ 600 Microliter Syringes, No Needle Included
Catalog No. p-7121963 Shop All Hamilton Company Products
Change view
Click to view available options
Volume (Metric):
2.5 μL
5 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
14-824-660 2.5 μL
14-824-661 5 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-824-660 Supplier Hamilton™ Supplier No. 763201
Only null left
Add to Cart
Add to Cart
The 600 series syringes are a half stroke version of the original 700 series syringes. This series of syringes is ideal for dispensing volumes from 0.25 μL up to 5 μL. The 600 series is often used in applications that require one handed operation because of the short stroke length and the reinforced plunger.
  • Half stroke version of the original icroliter 700 series syringes
  • Ideal for dispensing volumes from 0.25 μL up to 5 μL
  • Often used in applications that require one handed operation because of the short stroke length and the reinforced plunger
Specifications
Needle Gauge 22s G
Volume (Metric) 2.5 μL