Description
Combines features of both reusable and disposable respirators to provide effective protection. 3M™ Half Face Mask Respirator 6000 Series are economical, low-maintenance, simple-to-handle, and extremely lightweight.
- Probed reusable respirator assembly 6000 Series used for quantitative fit testing
- Probed facepiece allows for numerical measurement of the amount of leakage into the respirator and includes a probed facepiece and P100 filters
Compliance
NIOSH approved under 42 CFR 84 standard for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters; in Canada, CSA standard Z94.4 and applicable local government requirements must be met for approval.
Specifications
Specifications
| Product Type | Quantitative Fit Test Assembly |
| Certifications/Compliance | EN 140:1998, NIOSH 42 CFR Part 84 |
| For Use With (Application) | Conduct Quantitative Fit Tests on Respirator 6000 Series |
| For Use With (Equipment) | Conduct Quantitative Fit Tests on Respirator 6000 Series |
| Includes | Probed facepiece and P100 filter |
| Material | Thermoplastic Elastomer |
| Model | 6200 Q |
| Size | Medium |