Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYDRANAL™ - Water Standard 1.0, Standard for Karl Fischer titration (water content 1 mg/g = 0.1%), verified against NIST SRM 2890 & NMIJ CRM 4222, Solstice
BIOPHARMAGSA_VA

Catalog No. 6000682 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 Kit
40 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-006-82 40 mL
60-050-007 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-006-82 Supplier Solstice Supplier No. 348286X40ML
Add to Cart
Add to Cart

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

CAS 108-32-7, 100-66-3
Quantity 40 mL
Flash Point 43°C (109.4°F)
UN Number UN2222
Chemical Name or Material HYDRANAL™-Water Standard 1.0
Hazard Statement P210-P370 + P378
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.