Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Buffer solution pH 4.0 (20°C), With fungicide, citric acid / sodium hydroxide / sodium chloride solution, traceable to SRM from NIST, Solstice

Catalog No. 60046287 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
5 L
10 L
6 x 1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-287 6 x 1 L
60-024-47 500 mL
60-024-48 5 L
60-024-45 10 L
60-024-44 100 mL
60-024-46 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Alternative option to part number 33643
Catalog No. 60-046-287 Supplier Solstice Supplier No. 336436X1L
Add to Cart
Add to Cart
Alternative option to part number 33643

With fungicide, citric acid / sodium hydroxide / sodium chloride solution, traceable to SRM from NIST

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 100°C
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%, ≤2.0000%
CAS 18996-35-5, 144-33-2, 7647-14-5
Density 1.0 g/cm3
Quantity 6 x 1 L
Solution Type Buffer
Packaging Poly Bottle
Chemical Name or Material Buffer solution pH 4.0 (20°C), With fungicide, citric acid / sodium hydroxide / sodium chloride solution, traceable to SRM from NIST
ShelfLife 1260 days from date of manufacture
Recommended Storage Room Temp
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.