Learn More
601/602 Purgeable Halocarbons Kit, Restek™
SDP
Catalog No. 06729183 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
Catalog No. Quantity
06-729-183 1 mL
1 options
Catalog No. 06-729-183 Supplier Restek Corporation Supplier No. 30061
Contains 1 mL each of these mixtures.
30042: 502.2 Calibration Mix 1 (gases)
  • Bromomethane (74-83-9)
  • Chloroethane (75-00-3)
  • Chloromethane (74-87-3)
  • Dichlorodifluoromethane (CFC-12) (75-71-8)
  • Trichlorofluoromethane (CFC-11) (75-69-4)
  • Vinyl chloride (75-01-4)
30021: 624 Calibration Mix 2
  • Benzene (71-43-2)
  • Carbon tetrachloride (56-23-5)
  • Chlorobenzene (108-90-7)
  • 2-Chloroethyl vinyl ether (110-75-8)
  • Dibromochloromethane (124-48-1)
  • 1,1-Dichloroethane (75-34-3)
  • 1,1-Dichloroethene (75-35-4)
  • 1,2-Dichloropropane (78-87-5)
  • Methylene chloride (75-09-2)
  • Tetrachloroethene (127-18-4)
  • 1,1,2-Trichloroethane (79-00-5)
  • Trichloroethene (79-01-6)
30022: 624 Calibration Mix 3
  • Bromodichloromethane (75-27-4)
  • Bromoform (75-25-2)
  • Chloroform (67-66-3)
  • 1,2-Dichlorobenzene (95-50-1)
  • 1,3-Dichlorobenzene (541-73-1)
  • 1,4-Dichlorobenzene (106-46-7)
  • 1,2-Dichloroethane (107-06-2)
  • trans-1,2-Dichloroethene (156-60-5)
  • cis-1,3-Dichloropropene (10061-01-5)
  • trans-1,3-Dichloropropene (10061-02-6)
  • Ethylbenzene (100-41-4)
  • 1,1,2,2-Tetrachloroethane (79-34-5)
  • Toluene (108-88-3)
  • 1,1,1-Trichloroethane (71-55-6)
Specifications
Quantity 1 mL
ShelfLife 6 months