Learn More
Thermo Scientific 60ML CLEAR COLLECTION VIALS
Supplier:  Thermo Scientific 048784
Encompass
Collect solvent and analytes extracted from Thermo Scientific(TM) Dionex(TM) ASE(TM) 150/200/350 Accelerated Solvent Extractors using these collection vials. Placed in the vial tray at the lower right of the system, these vials are 60mL clear and amber glass, and are complete with caps and solvent-resistant (TFE) septa. Quantity 72
Catalog No. NC9337608